Assay Details
| Gene Symbol: LOC114115014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970705.1 | XP_027826506.1 | 2 - 3 | 106 | 111 | |
| Gene Symbol: LOC105606703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096122.2 | XP_014951608.1 | 2 - 3 | 133 | 111 | |
More Information
Set Membership: |
|

LOC114115014
Genomic Map