Assay Details
| Gene Symbol: STAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014248.4 | XP_004014297.1 | 5 - 6 | 601 | 65 | |
| XM_012188447.3 | XP_012043837.1 | 4 - 5 | 516 | 65 | ||
More Information
Set Membership: |
|

STAM
Genomic Map