Assay Details
| Gene Symbol: MAP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980307.1 | XP_027836108.1 | 2 - 3 | 254 | 70 | |
| XM_027980308.1 | XP_027836109.1 | 2 - 3 | 171 | 70 | ||
| XM_027980309.1 | XP_027836110.1 | 2 - 3 | 253 | 70 | ||
| XM_027980310.1 | XP_027836111.1 | 2 - 3 | 253 | 70 | ||
| XM_027980312.1 | XP_027836113.1 | 2 - 3 | 253 | 70 | ||
| XM_027980313.1 | XP_027836114.1 | 2 - 3 | 253 | 70 | ||
| XM_027980314.1 | XP_027836115.1 | 2 - 3 | 254 | 70 | ||
| XM_027980315.1 | XP_027836116.1 | 2 - 3 | 253 | 70 | ||
| XM_027980316.1 | XP_027836117.1 | 2 - 3 | 253 | 70 | ||
More Information
Set Membership: |
|

MAP9
Genomic Map