Assay Details
| Gene Symbol: FAM214B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964451.1 | XP_027820252.1 | 3 - 4 | 1431 | 54 | |
| XM_027964452.1 | XP_027820253.1 | 3 - 4 | 1302 | 54 | ||
| XM_027964453.1 | XP_027820254.1 | 3 - 4 | 1188 | 54 | ||
More Information
Set Membership: |
|

FAM214B
Genomic Map