Assay Details
| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980160.1 | XP_027835961.1 | 2 - 3 | 275 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTULINL| Gene Symbol: OTULINL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980160.1 | XP_027835961.1 | 2 - 3 | 275 | 81 | |