Assay Details
| Gene Symbol: LCA5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103821.3 | XP_011959211.2 | 7 - 8 | 1563 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LCA5L| Gene Symbol: LCA5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012103821.3 | XP_011959211.2 | 7 - 8 | 1563 | 79 | |
Set Membership: |
|