Assay Details
| Gene Symbol: TMEM45B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019547.4 | XP_004019596.1 | 5 - 6 | 902 | 59 | |
| XM_027959908.1 | XP_027815709.1 | 5 - 6 | 1007 | 59 | ||
| XM_027959909.1 | XP_027815710.1 | 5 - 6 | 908 | 59 | ||
More Information
Set Membership: |
|

TMEM45B
Genomic Map