Assay Details
| Gene Symbol: ZNF704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011760.4 | XP_004011809.1 | 6 - 7 | 1024 | 66 | |
| XM_027973082.1 | XP_027828883.1 | 6 - 7 | 1488 | 66 | ||
More Information
Set Membership: |
|

ZNF704
Genomic Map