Assay Details
| Gene Symbol: AP2M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003097.3 | XP_004003146.2 | 10 - 11 | 1249 | 66 | |
| XM_004003098.3 | XP_004003147.2 | 9 - 10 | 1209 | 66 | ||
| XM_027957753.1 | XP_027813554.1 | 11 - 12 | 1222 | 66 | ||
| XM_027957768.1 | XP_027813569.1 | 10 - 11 | 1216 | 66 | ||
More Information
Set Membership: |
|

AP2M1
Genomic Map