Assay Details
| Gene Symbol: GAREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103608.2 | XP_014959094.2 | 2 - 3 | 285 | 80 | |
| XM_027960788.1 | XP_027816589.1 | 2 - 3 | 285 | 80 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAREM1| Gene Symbol: GAREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103608.2 | XP_014959094.2 | 2 - 3 | 285 | 80 | |
| XM_027960788.1 | XP_027816589.1 | 2 - 3 | 285 | 80 | ||