Assay Details
| Gene Symbol: TP53BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013619.4 | XP_004013668.1 | 13 - 14 | 2886 | 61 | |
| XM_012187119.3 | XP_012042509.1 | 14 - 15 | 2795 | 61 | ||
| XM_027976293.1 | XP_027832094.1 | 13 - 14 | 2875 | 61 | ||
More Information
Set Membership: |
|

TP53BP2
Genomic Map