Assay Details
| Gene Symbol: LOC114112156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963732.1 | XP_027819533.1 | 10 - 11 | 2189 | 97 | |
| Gene Symbol: DHX57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966915.1 | XP_027822716.1 | 12 - 13 | 2481 | 97 | |
More Information
Set Membership: |
|

LOC114112156
Genomic Map