Assay Details
| Gene Symbol: LOC114112156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963732.1 | XP_027819533.1 | 4 - 5 | 1359 | 91 | |
| Gene Symbol: DHX57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966915.1 | XP_027822716.1 | 6 - 7 | 1651 | 91 | |
More Information
Set Membership: |
|

LOC114112156
Genomic Map