Assay Details
| Gene Symbol: LAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099527.2 | XP_014955013.1 | 1 - 2 | 565 | 69 | |
| XM_027976470.1 | XP_027832271.1 | 1 - 2 | 543 | 69 | ||
More Information
Set Membership: |
|

LAMP5
Genomic Map