Assay Details
| Gene Symbol: ENKUR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014226.4 | XP_004014275.1 | 3 - 4 | 924 | 82 | |
| XM_027976631.1 | XP_027832432.1 | 2 - 3 | 785 | 82 | ||
More Information
Set Membership: |
|

ENKUR
Genomic Map