Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012098057.3 | XP_011953447.2 | 1 - 2 | 569 | 80 | |
| XM_012098058.3 | XP_011953448.2 | 1 - 2 | 574 | 80 | ||
More Information
Set Membership: |
|

MTFP1
Genomic Map