Assay Details
| Gene Symbol: WASHC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097738.2 | XP_014953224.1 | 8 - 9 | 1267 | 65 | |
| XM_027972908.1 | XP_027828709.1 | 8 - 9 | 1267 | 65 | ||
More Information
Set Membership: |
|

WASHC5
Genomic Map