Assay Details
| Gene Symbol: LOC114112156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963732.1 | XP_027819533.1 | 5 - 6 | 1479 | 82 | |
| Gene Symbol: DHX57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966915.1 | XP_027822716.1 | 7 - 8 | 1771 | 82 | |
More Information
Set Membership: |
|

LOC114112156
Genomic Map