Assay Details
| Gene Symbol: AADACL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013803.3 | XP_004013852.3 | 1 - 2 | 1742 | 73 | |
| XM_027975883.1 | XP_027831684.1 | 4 - 5 | 734 | 73 | ||
| XM_027975884.1 | XP_027831685.1 | 3 - 4 | 424 | 73 | ||
| XM_027975885.1 | XP_027831686.1 | 5 - 6 | 856 | 73 | ||
| XM_027975886.1 | XP_027831687.1 | 2 - 3 | 224 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027975885.1, XM_027975886.1 transcripts.
Set Membership: |
|

AADACL3
Genomic Map