Assay Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014582.4 | XP_004014631.1 | 3 - 4 | 354 | 71 | |
| XM_004014583.4 | XP_004014632.1 | 4 - 5 | 455 | 71 | ||
| XM_012189222.3 | XP_012044612.1 | 4 - 5 | 1011 | 71 | ||
More Information
Set Membership: |
|

OSER1
Genomic Map