Assay Details
| Gene Symbol: MED20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958999.1 | XP_027814800.1 | 3 - 4 | 547 | 82 | |
| XM_027959000.1 | XP_027814801.1 | 3 - 4 | 511 | 82 | ||
More Information
Set Membership: |
|

MED20
Genomic Map