Assay Details
| Gene Symbol: LOC101120701 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018866.4 | XP_004018915.1 | 8 - 9 | 1333 | 71 | |
| XM_027958406.1 | XP_027814207.1 | 8 - 9 | 1334 | 71 | ||
More Information
Set Membership: |
|

LOC101120701
Genomic Map