Assay Details
| Gene Symbol: EEF2KMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104077.3 | XP_011959467.2 | 1 - 2 | 164 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EEF2KMT| Gene Symbol: EEF2KMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012104077.3 | XP_011959467.2 | 1 - 2 | 164 | 58 | |
Set Membership: |
|