Assay Details
| Gene Symbol: MPST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006728.4 | XP_004006777.1 | 2 - 3 | 703 | 76 | |
| XM_004006729.4 | XP_004006778.1 | 2 - 3 | 724 | 76 | ||
| XM_012175023.3 | XP_012030413.1 | 2 - 3 | 758 | 76 | ||
More Information
Set Membership: |
|

MPST
Genomic Map