Assay Details
| Gene Symbol: NUDT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101534.2 | XP_014957020.1 | 4 - 5 | 578 | 101 | |
| XM_027956315.1 | XP_027812116.1 | 4 - 5 | 578 | 101 | ||
| XM_027956316.1 | XP_027812117.1 | 4 - 5 | 571 | 101 | ||
| XM_027956317.1 | XP_027812118.1 | 2 - 3 | 73 | 101 | ||
More Information
Set Membership: |
|

NUDT6
Genomic Map