Assay Details
| Gene Symbol: ADAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980230.1 | XP_027836031.1 | 3 - 4 | 529 | 68 | |
| XM_027980231.1 | XP_027836032.1 | 3 - 4 | 529 | 68 | ||
More Information
Set Membership: |
|

ADAD1
Genomic Map