Assay Details
| Gene Symbol: LOC101103916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100463.2 | XP_014955949.1 | 1 - 2 | 73 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101103916| Gene Symbol: LOC101103916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100463.2 | XP_014955949.1 | 1 - 2 | 73 | 78 | |
Set Membership: |
|