Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182462.3 | XP_012037852.2 | 4 - 5 | 239 | 83 | |
| XM_015092270.2 | XP_014947756.2 | 3 - 4 | 1760 | 83 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map