Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961428.1 | XP_027817229.1 | 1 - 2 | 180 | 101 | |
| XM_027961429.1 | XP_027817230.1 | 1 - 2 | 180 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027961428.1, XM_027961429.1 transcripts.
Set Membership: |
|

ZNF500
Genomic Map