Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961428.1 | XP_027817229.1 | 6 - 7 | 1249 | 66 | |
| XM_027961429.1 | XP_027817230.1 | 6 - 7 | 1159 | 66 | ||
More Information
Set Membership: |
|

ZNF500
Genomic Map