Assay Details
| Gene Symbol: C8H6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972594.1 | XP_027828395.1 | 8 - 9 | 920 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8H6orf163| Gene Symbol: C8H6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972594.1 | XP_027828395.1 | 8 - 9 | 920 | 80 | |
Set Membership: |
|