Assay Details
| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960377.1 | XP_027816178.1 | 2 - 3 | 711 | 67 | |
| XM_027960378.1 | XP_027816179.1 | 3 - 4 | 749 | 67 | ||
More Information
Set Membership: |
|

DHX32
Genomic Map