Assay Details
| Gene Symbol: LRPAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971270.1 | XP_027827071.1 | 1 - 2 | 255 | 63 | |
| XM_027971271.1 | XP_027827072.1 | 1 - 2 | 296 | 63 | ||
More Information
Set Membership: |
|

LRPAP1
Genomic Map