Assay Details
| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963671.1 | XP_027819472.1 | 1 - 2 | 98 | 71 | |
| XM_027963672.1 | XP_027819473.1 | 1 - 2 | 92 | 71 | ||
| XM_027963673.1 | XP_027819474.1 | 1 - 2 | 98 | 71 | ||
| XM_027963674.1 | XP_027819475.1 | 1 - 2 | 98 | 71 | ||
More Information
Set Membership: |
|

MAP7D3
Genomic Map