Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012119208.2 | XP_011974598.1 | 3 - 4 | 487 | 68 | |
| XM_012119222.3 | XP_011974612.1 | 3 - 4 | 488 | 68 | ||
More Information
Set Membership: |
|

PEBP4
Genomic Map