Assay Details
| Gene Symbol: APEX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010390.3 | XP_004010439.2 | 4 - 5 | 588 | 65 | |
| XM_012181085.2 | XP_012036475.2 | 4 - 5 | 614 | 65 | ||
| XM_012181086.2 | XP_012036476.2 | 4 - 5 | 610 | 65 | ||
| XM_012181087.3 | XP_012036477.2 | 4 - 5 | 629 | 65 | ||
More Information
Set Membership: |
|

APEX1
Genomic Map