Assay Details
Gene Symbol: INO80E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020885.4 | XP_004020934.1 | 2 - 3 | 776 | 64 | |
XM_004020886.4 | XP_004020935.1 | 2 - 3 | 777 | 64 | ||
XM_012104365.3 | XP_011959755.1 | 2 - 3 | 318 | 64 | ||
XM_015104157.2 | XP_014959643.1 | 2 - 3 | 318 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
