Assay Details
| Gene Symbol: ECI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958630.1 | XP_027814431.1 | 5 - 6 | 628 | 64 | |
| XM_027958631.1 | XP_027814432.1 | 6 - 7 | 789 | 64 | ||
More Information
Set Membership: |
|

ECI2
Genomic Map