Assay Details
| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095060.3 | XP_011950450.3 | 5 - 6 | 850 | 51 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSK3A| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095060.3 | XP_011950450.3 | 5 - 6 | 850 | 51 | |
Set Membership: |
|