Assay Details
| Gene Symbol: RASSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959967.1 | XP_027815768.1 | 2 - 3 | 211 | 69 | |
| XM_027959968.1 | XP_027815769.1 | 2 - 3 | 232 | 69 | ||
More Information
Set Membership: |
|

RASSF7
Genomic Map