Assay Details
| Gene Symbol: XPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957820.1 | XP_027813621.1 | 7 - 8 | 927 | 61 | |
| XM_027957821.1 | XP_027813622.1 | 7 - 8 | 927 | 61 | ||
| XM_027957822.1 | XP_027813623.1 | 7 - 8 | 927 | 61 | ||
| XM_027957823.1 | XP_027813624.1 | 4 - 5 | 438 | 61 | ||
More Information
Set Membership: |
|

XPC
Genomic Map