Assay Details
| Gene Symbol: TNNI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976052.1 | XP_027831853.1 | 4 - 5 | 165 | 106 | |
| XM_027976053.1 | XP_027831854.1 | 4 - 5 | 166 | 106 | ||
| GenBank mRNA | KT218688.1 | - | 4 - 5 | 130 | 106 | |
| KT218689.1 | - | 4 - 5 | 166 | 106 | ||
More Information
Set Membership: |
|

TNNI1
Genomic Map