Assay Details
| Gene Symbol: TMBIM7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176317.3 | XP_012031707.1 | 8 - 9 | 839 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMBIM7P| Gene Symbol: TMBIM7P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176317.3 | XP_012031707.1 | 8 - 9 | 839 | 105 | |
Set Membership: |
|