Assay Details
| Gene Symbol: RIPPLY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012108635.3 | XP_011964025.2 | 2 - 3 | 175 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIPPLY3| Gene Symbol: RIPPLY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012108635.3 | XP_011964025.2 | 2 - 3 | 175 | 88 | |
Set Membership: |
|