Assay Details
| Gene Symbol: LOC101111528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965195.1 | XP_027820996.1 | 2 - 3 | 910 | 61 | |
| XM_027965196.1 | XP_027820997.1 | 2 - 3 | 921 | 61 | ||
More Information
Set Membership: |
|

LOC101111528
Genomic Map