Assay Details
| Gene Symbol: TAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015091961.2 | XP_014947447.2 | 3 - 4 | 3183 | 62 | |
| XM_027969765.1 | XP_027825566.1 | 4 - 5 | 1537 | 62 | ||
More Information
Set Membership: |
|

TAL1
Genomic Map