Assay Details
| Gene Symbol: OPN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104312.2 | XP_014959798.2 | 4 - 5 | 459 | 60 | |
| XM_027962232.1 | XP_027818033.1 | 3 - 4 | 423 | 60 | ||
More Information
Set Membership: |
|

OPN4
Genomic Map