Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974421.1 | XP_027830222.1 | 4 - 5 | 935 | 84 | |
| XM_027974422.1 | XP_027830223.1 | 6 - 7 | 1001 | 84 | ||
More Information
Set Membership: |
|

DBF4B
Genomic Map