Assay Details
| Gene Symbol: FAM131A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003087.3 | XP_004003136.2 | 4 - 5 | 733 | 103 | |
| XM_027957607.1 | XP_027813408.1 | 4 - 5 | 584 | 103 | ||
| XM_027957612.1 | XP_027813413.1 | 5 - 6 | 680 | 103 | ||
| XM_027957616.1 | XP_027813417.1 | 4 - 5 | 1524 | 103 | ||
| XM_027957623.1 | XP_027813424.1 | 3 - 4 | 2704 | 103 | ||
More Information
Set Membership: |
|

FAM131A
Genomic Map