Assay Details
| Gene Symbol: FGF9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973665.1 | XP_027829466.1 | 2 - 3 | 1054 | 82 | |
| GenBank mRNA | MH117879.1 | - | 2 - 3 | 593 | 82 | |
More Information
Set Membership: |
|

FGF9
Genomic Map